Gene Transcripts

Gene Symbol
Slc9b2
Entrez Gene ID
679958
Gene Name
solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2
Gene Aliases
Nhedc2
Chromosome Location
Chr. 2: 240486054 - 240519150 on Build Rnor_6.0
UniGene ID
Rn.198171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109385.2 NP_001102855.1 10 1352
GenBank mRNA BC166815.1 - 10 1352

View Full Product Details