Gene Transcripts

Gene Symbol
Pcdhb8
Entrez Gene ID
680020
Gene Name
protocadherin beta 8
Gene Aliases
Chromosome Location
Chr. 18: 30435119 - 30437458 on Build Rnor_6.0
UniGene ID
Rn.220357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014779.1 NP_001014779.1 1 202

View Full Product Details