Gene Transcripts

Gene Symbol
Gdpd4
Entrez Gene ID
680990
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases
Chromosome Location
Chr. 1: 162890482 - 162999849 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229796.2 XP_006229858.1 11 1090
XM_006229799.2 XP_006229861.1 11 1088

View Full Product Details