Gene Transcripts

Gene Symbol
Myeov2
Entrez Gene ID
681389
Gene Name
myeloma overexpressed 2
Gene Aliases
Chromosome Location
Chr. 9: 99813332 - 99818262 on Build Rnor_6.0
UniGene ID
Rn.14866
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109044.1 NP_001102514.1 3 387
GenBank mRNA DV717666.1 - 3 387
FQ212403.1 - 3 379
FQ221993.1 - 3 378
FQ223515.1 - 3 378

View Full Product Details