Gene Transcripts

Gene Symbol
WTAP
Entrez Gene ID
9589
Gene Name
Wilms tumor 1 associated protein
Gene Aliases
Mum2
Chromosome Location
Chr. 6: 159726680 - 159756319 on Build GRCh38
UniGene ID
Hs.446091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270531.1 NP_001257460.1 4, 5 355
NM_001270532.1 NP_001257461.1 4, 5 487
NM_001270533.1 NP_001257462.1 4, 5 355
NM_004906.4 NP_004897.2 4, 5 487
NM_152857.2 NP_690596.1 4, 5 355
NM_152858.2 NP_690597.1 4, 5 487
XM_017011514.1 XP_016867003.1 4, 5 325
XM_017011515.1 XP_016867004.1 2, 3 587
XM_017011516.1 XP_016867005.1 4, 5 325
GenBank mRNA AF374416.1 - 4, 5 368
AJ276706.1 - 3, 4 135
AK127822.1 - 4, 5 366
AK290854.1 - 4, 5 368
BC000383.2 - 4, 5 346
BC004432.1 - 4, 5 346
BC069192.1 - 4, 5 323 4653831
BE906617.1 - 4, 5 472
BG702882.1 - 4, 5 359
BQ948913.1 - 4, 5 293
BX387163.2 - 4, 5 337
CN284477.1 - 4, 5 317
D14661.1 - 4, 5 267
DA138013.1 - 4, 5 487
DA561768.1 - 4, 5 361

View Full Product Details