Gene Transcripts

Gene Symbol
Svep1
Entrez Gene ID
685899
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
Chromosome Location
Chr. 5: 75147245 - 75320871 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238194.2 XP_006238256.1 22, 23 5464

View Full Product Details