Gene Transcripts

Gene Symbol
Phemx
Entrez Gene ID
365390
Gene Name
pan hematopoietic expression
Gene Aliases
Chromosome Location
Chr. 1: 216189535 - 216206202 on Build Rnor_6.0
UniGene ID
Rn.154906
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728846.4 XP_002728892.3 2 2508
XM_006230915.2 XP_006230977.1 2 2508
XM_006230916.2 XP_006230978.1 2 2508
GenBank mRNA BC088266.1 - 2 63
BC105909.1 - 2 124
BC158837.1 - 1 48

View Full Product Details