Gene Transcripts

Gene Symbol
Mtx3
Entrez Gene ID
688905
Gene Name
metaxin 3
Gene Aliases
Chromosome Location
Chr. 2: 22427452 - 22440813 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749167.3 XP_003749215.1 3 230
XM_006231788.2 XP_006231850.1 3 230
XM_008760655.1 XP_008758877.1 3 230

View Full Product Details