Gene Transcripts

Gene Symbol
EMC2
Entrez Gene ID
9694
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
KIAA0103, TTC35
Chromosome Location
Chr. 8: 108443624 - 108486907 on Build GRCh38
UniGene ID
Hs.654351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK222615.1 - 7, 8 530
AK290974.1 - 7, 8 516
BC020753.1 - 7, 8 528 4809693
BC021667.1 - 7, 8 545 3682913
CR457402.1 - 7, 8 506
D14659.1 - 7, 8 512

View Full Product Details