Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142610.1 | NP_001136082.1 | 25 | 3283 | |
NM_014683.3 | NP_055498.3 | 25 | 3283 | ||
XM_011524087.2 | XP_011522389.1 | 21 | 2580 | ||
XM_017025424.1 | XP_016880913.1 | 25 | 3171 | ||
XM_017025425.1 | XP_016880914.1 | 25 | 3171 | ||
XM_017025426.1 | XP_016880915.1 | 25 | 3090 | ||
XM_017025427.1 | XP_016880916.1 | 23 | 2625 | ||
XM_017025428.1 | XP_016880917.1 | 23 | 2544 | ||
GenBank mRNA | AB014523.1 | - | 25 | 3274 | |
AK098316.1 | - | 24 | 2908 | ||
AK314345.1 | - | 25 | 2806 | ||
BC034988.1 | - | 26 | 2848 | 5268025 |