Gene Transcripts

Gene Symbol
Atp8b5p
Entrez Gene ID
691125
Gene Name
ATPase, class I, type 8B, member 5, pseudogene
Gene Aliases
Chromosome Location
Chr. 5: 58752558 - 58830617 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238129.2 XP_006238191.1 15, 16 1875
XM_008763657.1 XP_008761879.1 15, 16 1839
XM_008763658.1 XP_008761880.1 15, 16 1875

View Full Product Details