Gene Transcripts

Gene Symbol
Spint3
Entrez Gene ID
685136
Gene Name
serine peptidase inhibitor, Kunitz type, 3
Gene Aliases
Chromosome Location
Chr. 3: 161024739 - 161028925 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235628.2 XP_006235690.1 1, 2 1069

View Full Product Details