Gene Transcripts

Gene Symbol
Baiap2l2
Entrez Gene ID
685357
Gene Name
BAI1-associated protein 2-like 2
Gene Aliases
RGD1308786
Chromosome Location
Chr. 7: 120492366 - 120518803 on Build Rnor_6.0
UniGene ID
Rn.137549
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271220.1 NP_001258149.1 4 396

View Full Product Details