Gene Transcripts

Gene Symbol
Ggct
Entrez Gene ID
362368
Gene Name
gamma-glutamyl cyclotransferase
Gene Aliases
Ggctl1, RGD1304876, RGD1562590
Chromosome Location
Chr. 4: 85186069 - 85192857 on Build Rnor_6.0
UniGene ID
Rn.129060
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108629.2 NP_001102099.1 4 539
XM_008762899.1 XP_008761121.1 3 457
GenBank mRNA FM060344.1 - 4 477

View Full Product Details