Gene Transcripts

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
Chromosome Location
Chr. 2: 27370496 - 27377533 on Build GRCh38
UniGene ID
Hs.278569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267059.1 NP_001253988.1 8, 9 889
NM_001267060.1 NP_001253989.1 7, 8 850
NM_001267061.1 NP_001253990.1 8, 9 760
NM_014748.3 NP_055563.1 8, 9 925
NR_049782.1 8, 9 1050
NR_049783.1 9, 10 1023
NR_049784.1 9, 10 999
NR_049785.1 7, 8 932
NR_049786.1 8, 9 881
NR_049787.1 6, 7 732
XM_011533203.1 XP_011531505.1 7, 8 699
XM_017005405.1 XP_016860894.1 6, 7 869
GenBank mRNA AK129689.1 - 8, 9 809
AK222543.1 - 8, 9 808
AK293252.1 - 9, 10 899
AK293464.1 - 8, 9 741
AK295278.1 - 6, 7 611
AK297054.1 - 8, 9 934
AK298620.1 - 8, 9 760
AK298856.1 - 8, 9 762
AK298869.1 - 7, 8 711
AK300144.1 - 8, 9 772
AK300641.1 - 6, 7 603
AK307122.1 - 7, 8 808
AK308487.1 - 7, 8 811
AK309640.1 - 6, 7 853
AK316373.1 - 8, 9 704
BC002524.1 - 8, 9 786 3140610
BC002610.1 - 8, 9 775 3143104
BC014620.2 - 8, 9 786 3010648
BC050590.1 - 8, 9 780 6044270
BT007167.1 - 8, 9 677
CR457081.1 - 8, 9 677
D31764.1 - 8, 9 899

View Full Product Details