Gene Transcripts

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
Chromosome Location
Chr. 2: 27370496 - 27377533 on Build GRCh38
UniGene ID
Hs.278569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267059.1 NP_001253988.1 6 702
NM_001267060.1 NP_001253989.1 5 663
NM_001267061.1 NP_001253990.1 6 573
NM_014748.3 NP_055563.1 6 738
NR_049782.1 6 863
NR_049783.1 7 836
NR_049784.1 7 812
NR_049785.1 5 745
NR_049786.1 6 694
NR_049787.1 4 545
NR_049788.1 4 545
XM_011533203.1 XP_011531505.1 5 512
XM_017005405.1 XP_016860894.1 5 512
GenBank mRNA AK129689.1 - 6 622
AK222543.1 - 6 621
AK293252.1 - 7 712
AK293464.1 - 6 554
AK295278.1 - 4 424
AK297054.1 - 6 747
AK298620.1 - 6 573
AK298856.1 - 6 575
AK298869.1 - 5 524
AK300144.1 - 6 585
AK300641.1 - 4 416
AK307122.1 - 5 621
AK308487.1 - 5 624
AK309640.1 - 5 496
AK316373.1 - 6 517
BC002524.1 - 6 599 3140610
BC002610.1 - 6 588 3143104
BC014620.2 - 6 599 3010648
BC050590.1 - 6 593 6044270
BT007167.1 - 6 490
CR457081.1 - 6 490
D31764.1 - 6 712

View Full Product Details