Gene Transcripts

Gene Symbol
Lyrm7
Entrez Gene ID
686506
Gene Name
LYR motif containing 7
Gene Aliases
Chromosome Location
Chr. 10: 40183061 - 40201992 on Build Rnor_6.0
UniGene ID
Rn.34277
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134729.1 NP_001128201.1 4 208
GenBank mRNA BC168191.1 - 4 208

View Full Product Details