Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256864.1 | NP_001243793.1 | 10 | 1469 | |
NM_001256865.1 | NP_001243794.1 | 11 | 1417 | ||
NM_014787.3 | NP_055602.1 | 10 | 1304 | ||
GenBank mRNA | AB007942.1 | - | 10 | 1244 | |
AK296408.1 | - | 11 | 1371 | ||
BC051722.1 | - | 10 | 1253 | ||
BC109279.2 | - | 10 | 1402 | 40028812 | |
BC109280.1 | - | 10 | 1405 | 40028814 | |
BX647203.1 | - | 10 | 1458 |