Gene Transcripts
- Gene Symbol
- JAKMIP2
- Entrez Gene ID
- 9832
- Gene Name
- janus kinase and microtubule interacting protein 2
- Gene Aliases
- JAMIP2, NECC1
- Chromosome Location
- Chr. 5: 147585439 - 147782848 on Build GRCh38
- UniGene ID
- Hs.740461
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270934.1 |
NP_001257863.1
|
9, 10 |
1944 |
|
NM_001270941.1 |
NP_001257870.1
|
9, 10 |
1944 |
|
NM_001282282.1 |
NP_001269211.1
|
8, 9 |
1667 |
|
NM_014790.4 |
NP_055605.2
|
9, 10 |
1944 |
|
XM_017010097.1 |
XP_016865586.1
|
10, 11 |
1938 |
|
GenBank mRNA |
AB011127.1 |
- |
9, 10 |
1638 |
|
AF273057.1 |
- |
9, 10 |
1785 |
|
AK291280.1 |
- |
9, 10 |
1666 |
|
AK299724.1 |
- |
8, 9 |
1593 |
|
BC017354.2 |
- |
9, 10 |
1709 |
5020741 |
EF512550.1 |
- |
8, 9 |
1403 |
|
View Full Product Details