Gene Transcripts

Gene Symbol
Zbtb34
Entrez Gene ID
689174
Gene Name
zinc finger and BTB domain containing 34
Gene Aliases
Chromosome Location
Chr. 3: 12452705 - 12470563 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001069858.5 XP_001069858.2 2 2405
XM_006234021.2 XP_006234083.1 2 1434
XM_006234022.2 XP_006234084.1 2 2212

View Full Product Details