Gene Transcripts

Gene Symbol
Xlr3a
Entrez Gene ID
690041
Gene Name
X-linked lymphocyte-regulated 3A
Gene Aliases
Chromosome Location
Chr. 1: 147033272 - 147049664 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229630.2 XP_006229692.1 3 864
XM_008759622.1 XP_008757844.1 3 957

Gene Symbol
LOC100911047
Entrez Gene ID
100911047
Gene Name
X-linked lymphocyte-regulated protein 3A-like
Gene Aliases
Chromosome Location
Chr. 1: 147000994 - 147012434 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229628.2 XP_006229690.1 3 865
XM_008759621.1 XP_008757843.1 3 958

View Full Product Details