Gene Transcripts

Gene Symbol
Eif4a3
Entrez Gene ID
688288
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
eIF4A-III
Chromosome Location
Chr. 10: 108415201 - 108425195 on Build Rnor_6.0
UniGene ID
Rn.202617
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100158.2 NP_001093628.1 3, 4 368
GenBank mRNA BC105875.1 - 3, 4 313
CO559861.1 - 3, 4 368
FQ234161.1 - 2, 3 89

View Full Product Details