Gene Transcripts
- Gene Symbol
- ZC3H11A
- Entrez Gene ID
- 9877
- Gene Name
- zinc finger CCCH-type containing 11A
- Gene Aliases
- ZC3HDC11A
- Chromosome Location
- Chr. 1: 203795537 - 203854128 on Build GRCh38
- UniGene ID
- Hs.532399
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319238.1 |
NP_001306167.1
|
15, 16 |
2680 |
|
NM_001319239.1 |
NP_001306168.1
|
15, 16 |
2575 |
|
NM_014827.5 |
NP_055642.3
|
15, 16 |
2251 |
|
XM_005245643.2 |
XP_005245700.1
|
15, 16 |
2270 |
|
XM_005245644.3 |
XP_005245701.1
|
14, 15 |
1954 |
|
XM_006711673.2 |
XP_006711736.1
|
15, 16 |
2117 |
|
XM_011510199.1 |
XP_011508501.1
|
13, 14 |
3322 |
|
XM_011510200.1 |
XP_011508502.1
|
14, 15 |
2342 |
|
XM_011510201.1 |
XP_011508503.1
|
15, 16 |
2528 |
|
XM_011510202.1 |
XP_011508504.1
|
13, 14 |
2267 |
|
XM_017002956.1 |
XP_016858445.1
|
14, 15 |
3576 |
|
XM_017002957.1 |
XP_016858446.1
|
14, 15 |
3428 |
|
XM_017002958.1 |
XP_016858447.1
|
14, 15 |
3415 |
|
XM_017002959.1 |
XP_016858448.1
|
15, 16 |
2488 |
|
XM_017002960.1 |
XP_016858449.1
|
14, 15 |
3576 |
|
XM_017002961.1 |
XP_016858450.1
|
13, 14 |
3322 |
|
XM_017002962.1 |
XP_016858451.1
|
14, 15 |
2342 |
|
XM_017002963.1 |
XP_016858452.1
|
15, 16 |
2134 |
|
XM_017002964.1 |
XP_016858453.1
|
14, 15 |
3488 |
|
XM_017002965.1 |
XP_016858454.1
|
13, 14 |
2817 |
|
GenBank mRNA |
AB014563.1 |
- |
12, 13 |
1762 |
|
AK095858.1 |
- |
15, 16 |
2548 |
|
AK296983.1 |
- |
11, 12 |
1609 |
|
AK310470.1 |
- |
3, 4 |
644 |
|
BC014268.2 |
- |
15, 16 |
2089 |
3844154 |
BC038513.2 |
- |
15, 16 |
2406 |
|
BX649148.1 |
- |
12, 13 |
5867 |
|
CR627439.1 |
- |
14, 15 |
3272 |
|
CR627446.1 |
- |
12, 13 |
5375 |
|
View Full Product Details