Gene Transcripts

Gene Symbol
Fam19a4
Entrez Gene ID
689043
Gene Name
family with sequence similarity 19 (chemokine (C-C motif)-like), member A4
Gene Aliases
Chromosome Location
Chr. 4: 129196658 - 129430251 on Build Rnor_6.0
UniGene ID
Rn.158675
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134700.1 NP_001128172.1 6 1125
GenBank mRNA BC160856.1 - 6 1125

View Full Product Details