Gene Transcripts

Gene Symbol
Nbeal1
Entrez Gene ID
689253
Gene Name
neurobeachin-like 1
Gene Aliases
Als2cr16, Nbeal1-ps1
Chromosome Location
Chr. 9: 66951421 - 67113395 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767175.1 XP_008765397.1 5 891

View Full Product Details