Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005461.4 | NP_005452.2 | 1 | 1280 | |
GenBank mRNA | AF134157.1 | - | 1 | 963 | |
AK027324.1 | - | 1 | 898 | ||
BC028098.1 | - | 1 | 911 | 5242606 | |
BC036689.1 | - | 1 | 1281 | 5261053 |