Gene Transcripts
- Gene Symbol
- Usp12
- Entrez Gene ID
- 360763
- Gene Name
- ubiquitin specific peptidase 12
- Gene Aliases
- Chromosome Location
- Chr. 12: 10037516 - 10090545 on Build Rnor_6.0
- UniGene ID
- Rn.206008
- Species
- Rattus norvegicus
- Gene Symbol
- LOC100912059
- Entrez Gene ID
- 100912059
- Gene Name
- ubiquitin carboxyl-terminal hydrolase 12-like
- Gene Aliases
- Chromosome Location
- Chr. 12: 10965939 - 10967320 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1561481
- Entrez Gene ID
- 501659
- Gene Name
- similar to ubiquitin specific protease 12
- Gene Aliases
- Chromosome Location
- Chr. 16: 70081041 - 70082422 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_577055.5 |
XP_577055.4
|
2 |
874 |
|
View Full Product Details