Gene Transcripts

Gene Symbol
Rdh8
Entrez Gene ID
690953
Gene Name
retinol dehydrogenase 8 (all-trans)
Gene Aliases
Chromosome Location
Chr. 8: 21835103 - 21842374 on Build Rnor_6.0
UniGene ID
Rn.110636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168593.1 NP_001162065.1 4, 5 531

View Full Product Details