Gene Transcripts

Gene Symbol
Pla2g4f
Entrez Gene ID
691907
Gene Name
phospholipase A2, group IVF
Gene Aliases
Chromosome Location
Chr. 3: 112069461 - 112083963 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006234810.2 XP_006234872.1 20 2524
XM_008762122.1 XP_008760344.1 20 2521

View Full Product Details