Gene Transcripts

Gene Symbol
IRF7
Entrez Gene ID
3665
Gene Name
interferon regulatory factor 7
Gene Aliases
IMD39, IRF-7H, IRF7A, IRF7B, IRF7C, IRF7H
Chromosome Location
Chr. 11: 612555 - 615999 on Build GRCh38
UniGene ID
Hs.166120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001572.3 NP_001563.2 11 1897
NM_004029.2 NP_004020.1 10 1810
NM_004031.2 NP_004022.2 9 1976
XM_005252906.3 XP_005252963.1 10 1962
XM_005252907.3 XP_005252964.1 10 1959
XM_005252909.3 XP_005252966.1 9 1875
XM_011520066.2 XP_011518368.1 11 1873
XM_017017674.1 XP_016873163.1 4 775
GenBank mRNA AF076494.1 - 9 1901
AK303752.1 - 9 1708
BC136555.1 - 9 1612 9020545
KC477210.1 - 8 1889
U53830.1 - 10 1819
U53831.1 - 9 1732
U53832.1 - 9 1651
U73036.1 - 11 1836

View Full Product Details