Gene Transcripts

Gene Symbol
PTMS
Entrez Gene ID
5763
Gene Name
parathymosin
Gene Aliases
ParaT
Chromosome Location
Chr. 12: 6766139 - 6770952 on Build GRCh38
UniGene ID
Hs.504613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002824.4 NP_002815.3 5 837
XM_011520987.1 XP_011519289.1 5 1150
GenBank mRNA BC017025.1 - 5 556
BC128229.1 - 6 504
BE791815.1 - 2 287
M24398.1 - 5 804

View Full Product Details