Gene Transcripts

Gene Symbol
HIST3H3
Entrez Gene ID
8290
Gene Name
histone cluster 3, H3
Gene Aliases
H3.4, H3/g, H3FT, H3t
Chromosome Location
Chr. 1: 228424845 - 228425325 on Build GRCh38
UniGene ID
Hs.248171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003493.2 NP_003484.1 1 439
GenBank mRNA BC069079.1 - 1 439
BC101837.1 - 1 487 8069343
BC101839.1 - 1 487 8069345

View Full Product Details