Gene Transcripts

Gene Symbol
TRAPPC6A
Entrez Gene ID
79090
Gene Name
trafficking protein particle complex 6A
Gene Aliases
TRS33
Chromosome Location
Chr. 19: 45162928 - 45178302 on Build GRCh38
UniGene ID
Hs.466929
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270891.1 NP_001257820.1 6 737
NM_001270892.1 NP_001257821.1 5 711
NM_001270893.1 NP_001257822.1 5 669
NM_024108.2 NP_077013.1 6 779
XM_017027295.1 XP_016882784.1 6 875
GenBank mRNA AF161407.1 - 6 731
BC001907.1 - 6 763 3534111
BC004450.1 - 6 764 2820830

View Full Product Details