Gene Transcripts
- Gene Symbol
- CARD14
- Entrez Gene ID
- 79092
- Gene Name
- caspase recruitment domain family member 14
- Gene Aliases
- BIMP2, CARMA2, PRP, PSORS2, PSS1
- Chromosome Location
- Chr. 17: 80169992 - 80209331 on Build GRCh38
- UniGene ID
- Hs.675480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001257970.1 |
NP_001244899.1
|
6, 7 |
1134 |
|
NM_024110.4 |
NP_077015.2
|
6, 7 |
1134 |
|
NM_052819.2 |
NP_438170.1
|
3, 4 |
415 |
|
XM_011525212.1 |
XP_011523514.1
|
8, 9 |
1655 |
|
XM_011525213.1 |
XP_011523515.1
|
8, 9 |
1397 |
|
XM_011525215.1 |
XP_011523517.1
|
7, 8 |
1457 |
|
XM_011525216.1 |
XP_011523518.1
|
7, 8 |
1157 |
|
XM_011525217.1 |
XP_011523519.1
|
7, 8 |
1213 |
|
XM_011525218.2 |
XP_011523520.1
|
7, 8 |
1196 |
|
XM_017025057.1 |
XP_016880546.1
|
8, 9 |
1655 |
|
XM_017025058.1 |
XP_016880547.1
|
8, 9 |
1655 |
|
GenBank mRNA |
AF322642.1 |
- |
6, 7 |
1172 |
|
AK296762.1 |
- |
3, 4 |
355 |
|
AK309299.1 |
- |
2, 3 |
445 |
|
AY032927.1 |
- |
5, 6 |
969 |
|
BC001326.1 |
- |
3, 4 |
415 |
|
BC018142.1 |
- |
6, 7 |
1143 |
3847282 |
EU652409.1 |
- |
5, 6 |
966 |
|
View Full Product Details