Gene Transcripts

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
Chromosome Location
Chr. 17: 80169992 - 80209331 on Build GRCh38
UniGene ID
Hs.675480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257970.1 NP_001244899.1 10 1656
NM_024110.4 NP_077015.2 10 1656
NM_052819.2 NP_438170.1 7 937
NR_047566.1 11 1653
XM_011525212.1 XP_011523514.1 12 2177
XM_011525213.1 XP_011523515.1 12 1919
XM_011525215.1 XP_011523517.1 11 1979
XM_011525216.1 XP_011523518.1 11 1679
XM_011525217.1 XP_011523519.1 11 1735
XM_011525218.2 XP_011523520.1 11 1718
XM_017025057.1 XP_016880546.1 12 2177
XM_017025058.1 XP_016880547.1 12 2177
GenBank mRNA AF322642.1 - 10 1694
AK054723.1 - 2 372
AK091123.1 - 11 1611
AK296762.1 - 7 877
AK309299.1 - 6 967
AX746790.1 - 11 1611
AY032927.1 - 9 1491
BC001326.1 - 7 937
BC018142.1 - 10 1665 3847282
EU652409.1 - 9 1488

View Full Product Details