Gene Transcripts

Gene Symbol
CDRT15
Entrez Gene ID
146822
Gene Name
CMT1A duplicated region transcript 15
Gene Aliases
Chromosome Location
Chr. 17: 14235856 - 14236833 on Build GRCh38
UniGene ID
Hs.735172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007530.1 NP_001007531.1 1 71
GenBank mRNA BC146859.1 - 1 97

View Full Product Details