Gene Transcripts
- Gene Symbol
- FAM90A1
- Entrez Gene ID
- 55138
- Gene Name
- family with sequence similarity 90 member A1
- Gene Aliases
- Chromosome Location
- Chr. 12: 8221260 - 8227618 on Build GRCh38
- UniGene ID
- Hs.196086
- Species
- Homo sapiens
- Gene Symbol
- FAM90A25P
- Entrez Gene ID
- 389633
- Gene Name
- family with sequence similarity 90, member A7 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 8: 12414521 - 12418032 on Build GRCh38
- UniGene ID
- Hs.650464
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073395.1 |
|
4 |
1648 |
|
GenBank mRNA |
AL832996.1 |
- |
4 |
1647 |
|
- Gene Symbol
- FAM90A7P
- Entrez Gene ID
- 441317
- Gene Name
- putative protein FAM90A7
- Gene Aliases
- FAM90A7
- Chromosome Location
- Chr. 8: 7556138 - 7559104 on Build GRCh38
- UniGene ID
- Hs.715411
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046343.1 |
|
3 |
1593 |
|
View Full Product Details