Gene Transcripts
- Gene Symbol
- CBWD2
- Entrez Gene ID
- 150472
- Gene Name
- COBW domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 113437691 - 113496204 on Build GRCh38
- UniGene ID
- Hs.531330
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172003.3 |
NP_742000.1
|
7, 8 |
761 |
|
XM_005263600.2 |
XP_005263657.1
|
7, 8 |
761 |
|
XM_005263603.2 |
XP_005263660.1
|
8, 9 |
795 |
|
XM_005263605.3 |
XP_005263662.1
|
8, 9 |
796 |
|
XM_011510667.1 |
XP_011508969.1
|
7, 8 |
761 |
|
XM_011510668.2 |
XP_011508970.1
|
4, 5 |
321 |
|
XM_011510669.2 |
XP_011508971.1
|
8, 9 |
805 |
|
XM_017003407.1 |
XP_016858896.1
|
7, 8 |
761 |
|
XM_017003408.1 |
XP_016858897.1
|
9, 10 |
925 |
|
GenBank mRNA |
AF452722.1 |
- |
7, 8 |
617 |
|
AK225251.1 |
- |
8, 9 |
775 |
|
AK290072.1 |
- |
7, 8 |
761 |
|
AK303653.1 |
- |
8, 9 |
800 |
|
AY343910.1 |
- |
7, 8 |
646 |
|
BC120991.1 |
- |
7, 8 |
632 |
40116409 |
BC120992.1 |
- |
7, 8 |
632 |
40116411 |
- Gene Symbol
- CBWD1
- Entrez Gene ID
- 55871
- Gene Name
- COBW domain containing 1
- Gene Aliases
- COBP
- Chromosome Location
- Chr. 9: 121038 - 179083 on Build GRCh38
- UniGene ID
- Hs.531330
- Species
- Homo sapiens
- Gene Symbol
- CBWD3
- Entrez Gene ID
- 445571
- Gene Name
- COBW domain containing 3
- Gene Aliases
- bA561O23.1
- Chromosome Location
- Chr. 9: 68232437 - 68300035 on Build GRCh38
- UniGene ID
- Hs.645337
- Species
- Homo sapiens
- Gene Symbol
- CBWD5
- Entrez Gene ID
- 220869
- Gene Name
- COBW domain containing 5
- Gene Aliases
- DC36
- Chromosome Location
- Chr. 9: 65666374 - 65734041 on Build GRCh38
- UniGene ID
- Hs.355950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286835.1 |
NP_001273764.1
|
8, 9 |
814 |
|
XM_005272748.1 |
XP_005272805.1
|
7, 8 |
686 |
|
XM_005272749.1 |
XP_005272806.1
|
7, 8 |
686 |
|
XM_005272750.1 |
XP_005272807.1
|
6, 7 |
626 |
|
XM_006716914.1 |
XP_006716977.1
|
7, 8 |
686 |
|
XM_017014456.1 |
XP_016869945.1
|
8, 9 |
617 |
|
XM_017014458.1 |
XP_016869947.1
|
7, 8 |
686 |
|
XM_017014459.1 |
XP_016869948.1
|
9, 10 |
865 |
|
GenBank mRNA |
AK097639.1 |
- |
6, 7 |
1002 |
|
AK292585.1 |
- |
8, 9 |
754 |
|
BC067803.1 |
- |
8, 9 |
752 |
5294979 |
BC141856.1 |
- |
8, 9 |
752 |
|
- Gene Symbol
- CBWD6
- Entrez Gene ID
- 644019
- Gene Name
- COBW domain containing 6
- Gene Aliases
- Chromosome Location
- Chr. 9: 41131309 - 41198865 on Build GRCh38
- UniGene ID
- Hs.597638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001085457.1 |
NP_001078926.1
|
7, 8 |
689 |
|
XM_017015019.1 |
XP_016870508.1
|
6, 7 |
689 |
|
XM_017015020.1 |
XP_016870509.1
|
6, 7 |
689 |
|
XM_017015021.1 |
XP_016870510.1
|
6, 7 |
689 |
|
XM_017015023.1 |
XP_016870512.1
|
6, 7 |
689 |
|
XM_017015026.1 |
XP_016870515.1
|
6, 7 |
689 |
|
XM_017015027.1 |
XP_016870516.1
|
6, 7 |
689 |
|
XM_017015028.1 |
XP_016870517.1
|
7, 8 |
557 |
|
XM_017015029.1 |
XP_016870518.1
|
3, 4 |
508 |
|
XM_017015030.1 |
XP_016870519.1
|
6, 7 |
689 |
|
XM_017015031.1 |
XP_016870520.1
|
2, 3 |
200 |
|
XM_017015032.1 |
XP_016870521.1
|
6, 7 |
689 |
|
XM_017015034.1 |
XP_016870523.1
|
1, 2 |
63 |
|
XM_017015035.1 |
XP_016870524.1
|
7, 8 |
749 |
|
View Full Product Details