Gene Transcripts
- Gene Symbol
- MIER2
- Entrez Gene ID
- 54531
- Gene Name
- MIER family member 2
- Gene Aliases
- KIAA1193, Mi-er2
- Chromosome Location
- Chr. 19: 305573 - 344793 on Build GRCh38
- UniGene ID
- Hs.101891
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB033019.1 |
- |
|
3982 |
|
- Gene Symbol
- LOC101929060
- Entrez Gene ID
- 101929060
- Gene Name
- uncharacterized LOC101929060
- Gene Aliases
- Chromosome Location
- Chr. 19: 300658 - 304361 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details