Gene Transcripts
- Gene Symbol
- DPCR1
- Entrez Gene ID
- 135656
- Gene Name
- diffuse panbronchiolitis critical region 1
- Gene Aliases
- PBLT
- Chromosome Location
- Chr. 6: 30941000 - 30954221 on Build GRCh38
- UniGene ID
- Hs.631993
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080870.3 |
NP_543146.2
|
2 |
1412 |
|
View Full Product Details