Gene Transcripts

Gene Symbol
SLC17A2
Entrez Gene ID
10246
Gene Name
solute carrier family 17 member 2
Gene Aliases
NPT3
Chromosome Location
Chr. 6: 25912754 - 25930727 on Build GRCh38
UniGene ID
Hs.591802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286123.1 NP_001273052.1 3 382
NM_001286125.1 NP_001273054.1 3 382
NM_005835.3 NP_005826.1 4 832
XM_005248784.2 XP_005248841.1 4 832
XM_006714949.3 XP_006715012.1 4 569
XM_006714950.2 XP_006715013.1 3 459
XM_006714951.1 XP_006715014.1 4 832
XM_017010159.1 XP_016865648.1 3 722
XM_017010160.1 XP_016865649.1 4 827
GenBank mRNA BC104822.1 - 3 355
BC112033.1 - 3 355 8327501
BC143323.1 - 3 353
U90544.1 - 4 717

View Full Product Details