Gene Transcripts

Gene Symbol
SAP18
Entrez Gene ID
10284
Gene Name
Sin3A associated protein 18
Gene Aliases
2HOR0202, SAP18P
Chromosome Location
Chr. 13: 21140514 - 21149085 on Build GRCh38
UniGene ID
Hs.524899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005870.4 NP_005861.2 3 376
XM_011534868.2 XP_011533170.1 2 256
GenBank mRNA AF153608.1 - 3 314
AK311748.1 - 3 297
AY550970.1 - 3 296
BC030836.1 - 3 344 4794888
CR457035.1 - 3 280
DB171768.1 - 3 378
U78303.1 - 3 311
U96915.1 - 3 296

View Full Product Details