Gene Transcripts

Gene Symbol
Cryba1
Entrez Gene ID
12957
Gene Name
crystallin, beta A1
Gene Aliases
BA3/A1, Cryb
Chromosome Location
Chr. 11: 77718732 - 77725781 on Build GRCm38
UniGene ID
Mm.22830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009965.2 NP_034095.1 5 394
GenBank mRNA AJ239052.1 - 5 394
AK084145.1 - 2 971
BC107342.2 - 5 394 40055280

View Full Product Details