Gene Transcripts
- Gene Symbol
- Igh-VS107
- Entrez Gene ID
- 16065
- Gene Name
- immunoglobulin heavy chain (S107 family)
- Gene Aliases
- IgG, IgH, VhNZA6
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.359198
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC111444.1 |
- |
1 |
40 |
|
View Full Product Details