Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026666.3 | NP_080942.1 | 3 | 1012 | |
GenBank mRNA | AK019173.1 | - | 2 | 910 | |
AK020146.1 | - | 2 | 896 | ||
AK147571.1 | - | 3 | 1013 | ||
AK179093.1 | - | 1 | 198 | ||
BC098372.1 | - | 1 | 172 | 30363389 |