Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010745.2 | NP_034875.1 | 3, 4 | 449 | |
GenBank mRNA | AB007599.1 | - | 3, 4 | 449 | |
AK027929.1 | - | 3, 4 | 438 | ||
BC065783.1 | - | 3, 4 | 354 | ||
BC096414.1 | - | 3, 4 | 417 | 849991 |