Gene Transcripts

Gene Symbol
Vmn1r30
Entrez Gene ID
171195
Gene Name
vomeronasal 1 receptor 30
Gene Aliases
V1rc22, V1rc9
Chromosome Location
Chr. 6: 58434848 - 58521728 on Build GRCm38
UniGene ID
Mm.389764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134177.1 NP_598938.1 1 868
GenBank mRNA BC138075.1 - 1 917
BC138078.1 - 1 917 8861097

View Full Product Details