Gene Transcripts

Gene Symbol
Mt2
Entrez Gene ID
17750
Gene Name
metallothionein 2
Gene Aliases
AA409533, MT-II, Mt-2
Chromosome Location
Chr. 8: 94172618 - 94173567 on Build GRCm38
UniGene ID
Mm.147226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008630.2 NP_032656.1 3 387
GenBank mRNA AK002567.1 - 3 222
BC031758.1 - 3 204 2651471

View Full Product Details