Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146255.2 | NP_666367.2 | 10 | 1620 | |
GenBank mRNA | AK044586.1 | - | 10 | 1512 | |
AK044671.1 | - | 10 | 1550 | ||
BC030400.1 | - | 10 | 1631 | 5363389 | |
BC054455.1 | - | 2 | 508 |