Gene Transcripts

Gene Symbol
Fam19a4
Entrez Gene ID
320701
Gene Name
family with sequence similarity 19, member A4
Gene Aliases
C130034I18Rik, Fam19a4 Tafa-4, Tafa-4
Chromosome Location
Chr. 6: 96831206 - 97080124 on Build GRCm38
UniGene ID
Mm.131700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177233.5 NP_796207.2 6 924
GenBank mRNA AK081531.1 - 6 908
AK149355.1 - 6 927
AY325123.1 - 6 707
BC089490.1 - 6 884 30504398

View Full Product Details